EMAIL THIS PAGE TO A FRIEND
Journal of veterinary pharmacology and therapeutics
Plasma and tissue pharmacokinetics of danofloxacin in healthy and in experimentally infected chickens with Pasteurella multocida.
PMID 21219355