EMAIL THIS PAGE TO A FRIEND
Journal of food protection
Inactivation of Bacillus amyloliquefaciens spores by a combination of sucrose laurate and pressure-assisted thermal processing.
PMID 21219716
Abstract
The aim of this research was to study the effect of sucrose laurate ester (SL) on enhancing pressure-assisted thermal processing (PATP) inactivation of Bacillus amyloliquefaciens Fad 82 spores. B. amyloliquefaciens spores (∼10⁸ CFU/ml) were suspended in deionized water, solutions of 0.1, 0.5, and 1.0% SL, and mashed carrots without or with 1% SL. Samples were treated at 700 MPa and 105°C for 0 (come-up time), 1, 2, and 5 min and analyzed by pour-plating and most-probable-number techniques. Heat shock (80°C, 10 min) was applied to untreated and treated samples to study the germination rates. Results were also compared against samples treated by high pressure processing (700 MPa, 35°C) and thermal processing (105°C, 0.1 MPa). Among the combinations tested, SL at concentrations of 1.0% showed the best synergistic effect against spores of B. amyloliquefaciens when combined with PATP treatments. In the case of high pressure and thermal processing treatments, SL did not enhance spore inactivation at the conditions tested. These results suggest that SL is a promising antimicrobial compound that can help reduce the severity of PATP treatments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
84110
Sucrose monolaurate, BioXtra, ≥97.0% (TLC)
C24H44O12