EMAIL THIS PAGE TO A FRIEND
Neuron
SIK2 is a key regulator for neuronal survival after ischemia via TORC1-CREB.
PMID 21220102
Abstract
The cAMP responsive element-binding protein (CREB) functions in a broad array of biological and pathophysiological processes. We found that salt-inducible kinase 2 (SIK2) was abundantly expressed in neurons and suppressed CREB-mediated gene expression after oxygen-glucose deprivation (OGD). OGD induced the degradation of SIK2 protein concomitantly with the dephosphorylation of the CREB-specific coactivator transducer of regulated CREB activity 1 (TORC1), resulting in the activation of CREB and its downstream gene targets. Ca(2+)/calmodulin-dependent protein kinase I/IV are capable of phosphorylating SIK2 at Thr484, resulting in SIK2 degradation in cortical neurons. Neuronal survival after OGD was significantly increased in neurons isolated from sik2(-/-) mice, and ischemic neuronal injury was significantly reduced in the brains of sik2(-)(/-) mice subjected to transient focal ischemia. These findings suggest that SIK2 plays critical roles in neuronal survival, is modulated by CaMK I/IV, and regulates CREB via TORC1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500955
Anti-SIK2 antibody produced in rabbit, affinity isolated antibody