EMAIL THIS PAGE TO A FRIEND
Nanoscale
Probing the role of aromaticity in the design of dipeptide based nanostructures.
PMID 21221462
Abstract
Self-assembly of peptide into nanostructures is believed to be stabilized primarily by aromatic interactions. Using a minimalistic approach, we probed the importance of aromatic interactions in the self-assembly of simple model dipeptides. Our results suggest that aromaticity may not be absolutely essential for self-assembly, even though it tends to provide directionality to the assembly. We found that peptides containing cyclic/linear side chain hydrophobic residues were also capable of forming stable self-assemblies that are stabilized by hydrophobic interactions. Our observations will find relevance in the design of small peptide based nanoparticles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4126
Phe-Phe
C18H20N2O3 · xH2O