EMAIL THIS PAGE TO A FRIEND
MMW Fortschritte der Medizin
[Musculoskeletal pain. Pain relief with muscle relaxant].
PMID 21222334
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F8927
Flupirtine maleate salt, ≥98% (HPLC)
C15H17FN4O2 · C4H4O4