EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
An approach toward homocalystegines and silyl-homocalystegines. acid-mediated migrations of acetates in seven-membered ring systems.
PMID 21222434
Abstract
A short access to homocalystegine analogues silylated at C7 is described. The synthesis involves the desymmetrization of a (phenyldimethylsilyl)methylcycloheptatriene using osmium-mediated dihydroxylation, followed by the diol protection and a cycloaddition involving the remaining diene moiety and an acylnitroso reagent. Additions of the osmium and acylnitroso reagents were shown, through X-ray diffraction studies of the resulting major isomers, to occur anti and syn, respectively, relative to the SiCH(2) substituent. N-O bond cleavage on the resulting cycloadduct then produces the aminopolyol having a silylmethyl substituent. Oxidation of the C-Si bond also afforded an access to unusual amino-heptitols having five contiguous stereogenic centers. In the course of this work, we finally observed a unusual rearrangement taking place on cycloheptanone 18 substituted by two acetyl groups and a neighboring Boc-protected amine. A profound reorganization of the substituents on the seven-membered ring effectively took place under acidic conditions (TFA) leading to the thermodynamically more stable homocalystegine-type compound. DFT calculations of the conformational energy of isomeric silyl homocalystegines indicated that the product observed upon the acid-mediated rearrangement was the most stable of a series of analogues with various distributions of substituents along the seven-membered ring backbone. A tentative mechanism is proposed to rationalize the acetate migrations and inversions of the stereochemistry at various stereocenters.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C99000
Cycloheptanone, 99%
C7H12O
164623
Tropylium tetrafluoroborate, 97%
C7H7BF4