EMAIL THIS PAGE TO A FRIEND
Xenobiotica; the fate of foreign compounds in biological systems
The effect of the tert-butylquinone metabolite of butylated hydroxyanisole on cytochrome P-450 monooxygenase activity.
PMID 2122606
Abstract
1. The t-butylquinone metabolite of BHA was shown to redox cycle with NADPH-cytochrome P-450 reductase leading to enhanced NADPH-oxidase activity for both the purified and liver microsome-bound flavoprotein. Likewise, addition of t-butylquinone (20-100 microM) strikingly inhibited electron transfer from the flavoprotein reductase to cytochrome P-450 of liver microsomes from phenobarbital-treated rats. 2. When the effect of t-butylquinone on metabolism of biphenyl was evaluated with liver microsomal fractions or isolated hepatocytes, t-butylquinone was less effective as an inhibitor then BHA alone or vitamin K3 (menadione). Addition of dicoumarol had little or no effect on the inhibitory potency of either t-butylquinone or vitamin K3 in isolated hepatocytes. 3. t-Butylquinone was not an effective reductant for exogenous oxidants, such as cytochrome c, in the presence of purified, cytosolic NAD(P)H-quinone oxidoreductase (DT-diaphorase). This property is most probably due to the lower rate of reoxidation of t-butylquinone by molecular oxygen, relative to vitamin K3 (menadione).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
429074
2-tert-Butyl-1,4-benzoquinone, 98%
C10H12O2