EMAIL THIS PAGE TO A FRIEND
MMW Fortschritte der Medizin
[Vertigo. Evidence based therapy of dizziness].
PMID 21226379
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5270
Cinnarizine, powder
C26H28N2
C2180000
Cinnarizine, European Pharmacopoeia (EP) Reference Standard
C26H28N2
D2396
Dimenhydrinate
C17H21NO · C7H7ClN4O2