EMAIL THIS PAGE TO A FRIEND
Acta poloniae pharmaceutica
5-Nitro-2-pyridyl-1-thioglucosides: application in synthesis of analogues of glycosyltransferases natural substrates.
PMID 21229881
Abstract
5-Nitro-2-pyridyl-1-thioglucosides were used in synthesis of complex uridine derivatives (13-16) in two different sequences of reactions. In one route, the first step was glycosylation of selectively protected 5-nitro-2-pyridyl-1-thioglucoside 1 with two different glycosyl donors (5 or 6), next, the nitro group in aglycone of obtained disaccharides 7 or 8 was reduced and then obtained products 9 or 10 were condensed with uridine derivatives 3 or 4 using DMT-MM as condensing agent under microwave irradiation. In the second route, condensation and glycosylation reactions were applied in reverse order. As it turned up, a sequence of reactions affected the yield of final glycoconjugates 13-16 and depended on the type of uridine derivatives used.