EMAIL THIS PAGE TO A FRIEND
Molecular biology of the cell
The Ccr4a (CNOT6) and Ccr4b (CNOT6L) deadenylase subunits of the human Ccr4-Not complex contribute to the prevention of cell death and senescence.
PMID 21233283
Abstract
A key step in cytoplasmic mRNA degradation is the shortening of the poly(A) tail, which involves several deadenylase enzymes. Relatively little is known about the importance of these enzymes for the cellular physiology. Here we focused on the role of the highly similar Ccr4a (CNOT6) and Ccr4b (CNOT6L) deadenylase subunits of the Ccr4-Not complex. In addition to a role in cell proliferation, Ccr4a and Ccr4b play a role in cell survival, in contrast to the Caf1a (CNOT7) and Caf1b (CNOT8) deadenylase subunits or the CNOT1 and CNOT3 noncatalytic subunits of the Ccr4-Not complex. Underscoring the differential contributions of the deadenylase subunits, we found that knockdown of Caf1a/Caf1b or Ccr4a/Ccr4b differentially affects the formation of cytoplasmic foci by processing-body components. Furthermore, we demonstrated that the amino-terminal leucine-rich repeat (LRR) domain of Ccr4b influenced its subcellular localization but was not required for the deadenylase activity of Ccr4b. Moreover, overexpression of Ccr4b lacking the LRR domain interfered with cell cycle progression but not with cell viability. Finally, gene expression profiling indicated that distinct gene sets are regulated by Caf1a/Caf1b and Ccr4a/Ccr4b and identified Ccr4a/Ccr4b as a key regulator of insulin-like growth factor-binding protein 5, which mediates cell cycle arrest and senescence via a p53-dependent pathway.