EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Effect of selenium-containing products on antioxidant enzyme activity in the kidneys, liver, and blood of guinea pigs.
PMID 21234431
Abstract
The antioxidant system of some tissues in guinea pigs with no selenium deficiency was studied after treatment with selenium-containing products. Activity of a selenium-containing enzyme glutathione peroxidase was significantly reduced in the kidneys and blood. The kidneys were characterized by low activity of glutathione reductase and high activity of SOD. These features illustrate the development of oxidative stress in the kidneys.