EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Highly diastereo- and enantio-selective epoxidation of secondary allylic alcohols catalyzed by styrene monooxygenase.
PMID 21234475
Abstract
Enantiomerically enriched glycidol derivatives with contiguous stereogenic centers were obtained in a highly diastereo- and enantio-selective epoxidation catalyzed with the styrene monooxygenase StyAB2.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
480819
(R)-(+)-Glycidol, 97%, optical purity ee: 98% (GLC)
C3H6O2
G5809
Glycidol, 96%
C3H6O2