EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Picosecond X-ray absorption measurements of the ligand substitution dynamics of Fe(CO)5 in ethanol.
PMID 21234500
Abstract
Ultrafast X-ray absorption near edge spectroscopy has been carried out for photo excited iron pentacarbonyl in ethanol with 2 picosecond resolution. A temporal resolution limited dissociation process was observed, followed by the formation of the mono-substituted complex Fe(CO)(4)EtOH within a few tens of picoseconds. The measurements have been carried out with a newly developed X-ray absorption instrument at station 7 ID-C of the Advanced Photon Source. The results show that single picosecond temporal resolution can be achieved at a synchrotron beam line.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
481718
Iron(0) pentacarbonyl, >99.99% trace metals basis
C5FeO5
195731
Iron(0) pentacarbonyl
C5FeO5