EMAIL THIS PAGE TO A FRIEND
Veterinary parasitology
Quantifying fenbendazole and its metabolites in self-medicating wild red grouse Lagopus lagopus scoticus using an HPLC-MS-MS approach.
PMID 21236579
Abstract
On red grouse estates in the UK the nematode parasite Trichostrongylus tenuis is often controlled by application of grit medicated with the anthelmintic fenbendazole (FBZ). To date, assessment of the efficacy has been inhibited by the inability to quantify uptake of FBZ by the birds. We have developed a simple and sensitive HPLC-MS-MS method for detecting and quantifying FBZ and its metabolites from a 300 mg sample of red grouse liver. This method could be used to improve the efficacy of medicated grit treatment by allowing the identification of conditions and application methods that optimize the uptake of FBZ. With the necessary modifications, our method will also be applicable to other wildlife species where self-medication is used for parasite control.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5396
Fenbendazole, ≥98%
C15H13N3O2S
35032
Fenbendazole, VETRANAL, analytical standard
C15H13N3O2S