EMAIL THIS PAGE TO A FRIEND
Journal of plant physiology
Early senescence induced by 2-3H-benzoxazolinone (BOA) in Arabidopsis thaliana.
PMID 21237530
Abstract
Measurements of chlorophyll a fluorescence, nutrient and trace elements, total protein content and malonyldialdehyde in leaves of Arabidopsis thaliana between 1 and 192 h after treatment with 0, 1 or 3 mM 2-3H-benzoxazolinone (BOA), together with imaging of chlorophyll a fluorescence and of the distributions of hydrogen peroxide and superoxide anion, suggested that the primary phytotoxic action of BOA is the induction of premature senescence, and that oxidative stress is a secondary effect that sets in a day or two later.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
157058
2-Benzoxazolinone, 98%
C7H5NO2