EMAIL THIS PAGE TO A FRIEND
Human & experimental toxicology
Anti-hepatotoxic effects of 3,4-methylenedioxyphenol and N-acetylcysteine in acutely acetaminophen-overdosed mice.
PMID 21239480
Abstract
3,4-Methylenedioxyphenol (sesamol) is effective against acetaminophen-induced liver injury in rats. Whether sesamol's anti-hepatotoxic effect is comparable to that of N-acetylcysteine has never been studied. We investigated the anti-hepatotoxic effects of sesamol and N-acetylcysteine on acetaminophen-induced hepatotoxicity in mice. Equimolar doses (1 mmol/kg) of sesamol and N-acetylcysteine significantly inhibited acetaminophen (300 mg/kg)-increased serum aspartate transaminase and alanine transaminase levels 6 h post-administration. Sesamol and N-acetylcysteine maintained hepatic glutathione levels and inhibited lipid peroxidation. Moreover, the combination of sesamol and N-acetylcysteine antagonistically inhibited sesamol's protection against acetaminophen-induced liver injury. We conclude that the protective effect of sesamol against acetaminophen-induced liver damage is comparable to that of N-acetylcysteine by maintaining glutathione levels and inhibiting lipid peroxidation in mice.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S3003
Sesamol, 98%
C7H6O3