EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Homopolymerization of ethylene, 1-hexene, styrene and copolymerization of styrene with 1,3-cyclohexadiene using (η⁵-tetramethylcyclopentadienyl)dimethylsilyl(N-Ar')amido-TiCl₂/MAO (Ar'=6-(2-(diethylboryl)phenyl)pyrid-2-yl, biphen-3-yl).
PMID 21240147
Abstract
The propensity of a half-sandwich (η⁵-tetramethylcyclopentadienyl) dimethylsilylamido Ti(IV)-based catalyst bearing an auxiliary diethylboryl-protected pyridyl moiety (Ti-8), activated by methylaluminoxane (MAO) to homopolymerize α-olefins such as ethylene, 1-hexene and styrene as well as to copolymerize styrene with 1,3-cyclo-hexadiene is described. The reactivity of Ti-8 was investigated in comparison to a 6-(2-(diethylboryl)phenyl)pyrid-2-yl-free analogue (Ti-3).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C100005
1,3-Cyclohexadiene, contains 0.05% BHT as inhibitor, 97%
C6H8