EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Role of the brain dopaminergic and opioid system in the regulation of "child's" (maternal bonding) behavior of newborn albino rats.
PMID 21240335
Abstract
Administration of D(2) receptor antagonist clebopride in a dose not affecting locomotor activity was followed by a decrease in maternal bonding behavior of 10-day-old and 15-day-old albino rat pups. D(1) receptor antagonist SCH23390 had a stimulatory effect only on the behavior of 10-day-old newborns. Opioid peptide β-casomorphin-7 abolished the effect of clebopride in rat pups of the older age group.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8289
Clebopride maleate salt, ~95%
C20H24ClN3O2 · C4H4O4