EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Modulation of reparative regeneration and CD117 expression by liver cells after partial hepatectomy in mice.
PMID 21240352
Abstract
The effects of 3-aminophthalhydrazide and carrageenan on reparative regeneration and expression of CD117 by liver cells after partial hepatectomy were studied in mice. 3-Aminophthalhydrazide stimulated regeneration of the liver and increased the count of CD117(+) hepatocytes. By contrast, carrageenan inhibited liver reparation and CD117 expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8264
4-Aminophthalhydrazide
C8H7N3O2