EMAIL THIS PAGE TO A FRIEND
British journal of cancer
Enhanced ZAG production by subcutaneous adipose tissue is linked to weight loss in gastrointestinal cancer patients.
PMID 21245862
Abstract
Profound loss of adipose tissue is a hallmark of cancer cachexia. Zinc-α2-glycoprotein (ZAG), a recently identified adipokine, is suggested as a candidate in lipid catabolism. In the first study, eight weight-stable and 17 cachectic cancer patients (weight loss 5% in previous 6 months) were recruited. Zinc-α2-glycoprotein mRNA and protein expression were assessed in subcutaneous adipose tissue (SAT), subcutaneous adipose tissue morphology was examined and serum ZAG concentrations were quantified. In the second cohort, ZAG release by SAT was determined in 18 weight-stable and 15 cachectic cancer patients. The effect of ZAG on lipolysis was evaluated in vitro. Subcutaneous adipose tissue remodelling in cancer cachexia was evident through shrunken adipocytes with increased fibrosis. In cachectic cancer patients, ZAG mRNA was upregulated (2.7-fold, P=0.028) while leptin mRNA decreased (2.2-fold, P=0.018); serum ZAG levels were found to be unaffected. Zinc-α2-glycoprotein mRNA correlated positively with weight loss (r=0.51, P=0.01) and serum glycerol levels (r=0.57, P=0.003). Zinc-α2-glycoprotein release by SAT was also elevated in cachectic patients (1.5-fold, P=0.024) and correlated with weight loss (r=0.50, P=0.003). Recombinant ZAG stimulated lipolysis in human adipocytes. Zinc-α2-glycoprotein expression and secretion by adipose tissue is enhanced in cachectic cancer patients. Given its lipid-mobilising effect, ZAG may contribute to adipose atrophy associated with cancer cachexia in human beings.