EMAIL THIS PAGE TO A FRIEND
British journal of pharmacology
Nicorandil ameliorates ischaemia-reperfusion injury in the rat kidney.
PMID 21250976
Abstract
Nicorandil, an ATP-sensitive potassium (K(ATP) ) channel opener and nitric oxide donor, is used in the treatment of angina and acute heart failure. Here we investigated the effects of two K(ATP) channel openers, nicorandil and cromakalim on ischaemia reperfusion (I-R) injury in the kidney. Right nephrectomy was performed in 8-week-old male Sprague-Dawley rats and they were then divided into six groups: control group; I-R, including 30 min of left renal ischaemia followed by 24 h of reperfusion; I-R groups plus nicorandil 3 or 10 mg·kg⁻¹ i.p.; and I-R groups plus cromakalim 100 or 300 µg·kg⁻¹ i.p. After reperfusion, renal function was estimated by serum creatinine (SCr), urinary albumin:creatinine ratio (ACR) and urinary β2-microglobulin (β2-MG). Levels of K(ATP) channel subtypes were investigated by Western blot. Kidney sections were stained for 4-hydroxy-2-nonenal and 8-hydroxy-2'-deoxyguanosine. Renal I-R induced significant increases in SCr, ACR and β2-MG levels compared with the control animals. Treatment with K(ATP) channel openers reduced urinary β2-MG levels, raised by I-R. Both K(IR) 6.1 and K(IR) 6.2 channels were expressed. Expression of K(IR) 6.2 channels in the I-R group was lower than in the control group, which was restored to normal by treatment with K(ATP) channel openers. Histologically, severe acute tubular damage was observed in the I-R kidney and this damage was ameliorated by K(ATP) channel openers, dose-dependently. ATP-sensitive potassium channel openers protected against proximal tubule damage after I-R injury. Nicorandil could represent a powerful additional component in the treatment of patients undergoing partial nephrectomy or renal transplantation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1055
Cromakalim
C16H18N2O3