EMAIL THIS PAGE TO A FRIEND
Chemical biology & drug design
Synthesis of 2-hydrazolyl-4-thiazolidinones based on multicomponent reactions and biological evaluation against Trypanosoma Cruzi.
PMID 21251233
Abstract
A series of 18 novel 2-hydrazolyl-4-thiazolidinones-5-carboxylic acids, amides and 5,6-α,β-unsaturated esters were synthesized, and their in vitro activity on cruzipain and T. cruzi epimastigotes was determined. Some agents show activity at 37 μm concentration in the enzyme assay. Computational tools and docking were used to correlate the biological response with the physicochemical parameters of the compounds and their cruzipain inhibitory effects.