EMAIL THIS PAGE TO A FRIEND
International Journal of Gynaecology and Obstetrics: the Official Organ of the International Federation of Gynaecology and Obstetrics
Accuracy of visual inspection with acetic acid for cervical cancer screening.
PMID 21257169