EMAIL THIS PAGE TO A FRIEND
The Veterinary record
Use of monepantel.
PMID 21257429
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5802
Aminoacetonitrile, ≥98%
C2H4N2