EMAIL THIS PAGE TO A FRIEND
The Veterinary record
Treatment of diarrhoea in cats caused by Tritrichomonas foetus.
PMID 21257565
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R7635
Ronidazole, analytical standard
C6H8N4O4