EMAIL THIS PAGE TO A FRIEND
Journal of inherited metabolic disease
The sodium-dependent di- and tricarboxylate transporter, NaCT, is not responsible for the uptake of D-, L-2-hydroxyglutarate and 3-hydroxyglutarate into neurons.
PMID 21264516
Abstract
Concentrations of glutarate (GA) and its derivatives such as 3-hydroxyglutarate (3OHGA), D- (D-2OHGA) and L-2-hydroxyglutarate (L-2OHGA) are increased in plasma, cerebrospinal fluid (CSF) and urine of patients suffering from different forms of organic acidurias. It has been proposed that these derivatives cause neuronal damage in these patients, leading to dystonic and dyskinetic movement disorders. We have recently shown that these compounds are eliminated by the kidneys via the human organic anion transporters, OAT1 and OAT4, and the sodium-dependent dicarboxylate transporter 3, NaDC3. In neurons, where most of the damage occurs, a sodium-dependent citrate transporter, NaCT, has been identified. Therefore, we investigated the impact of GA derivatives on hNaCT by two-electrode voltage clamp and tracer uptake studies. None of these compounds induced substrate-associated currents in hNaCT-expressing Xenopus laevis oocytes nor did GA derivatives inhibit the uptake of citrate, the prototypical substrate of hNaCT. In contrast, D- and L-2OHGA, but not 3OHGA, showed affinities to NaDC3, indicating that D- and L-2OHGA impair the uptake of dicarboxylates into astrocytes thereby possibly interfering with their feeding of tricarboxylic acid cycle intermediates to neurons.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
89147
Glutaric acid, certified reference material, TraceCERT®
C5H8O4