EMAIL THIS PAGE TO A FRIEND
Chudoku kenkyu : Chudoku Kenkyukai jun kikanshi = The Japanese journal of toxicology
[Acute tebufenpyrad and tolfenpyrad poisoning in humans].
PMID 21268826
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46438
Tebufenpyrad, PESTANAL®, analytical standard
C18H24ClN3O