EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Contact dermatitis and secondary systemic allergy to dimethindene maleate.
PMID 21272026
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2205000 Dimetindene maleate, European Pharmacopoeia (EP) Reference Standard
C20H24N2 · C4H4O4