EMAIL THIS PAGE TO A FRIEND
Brain research bulletin
TPH2 in the ventral tegmental area of the male rat brain.
PMID 21272616
Abstract
This study surveyed the distribution of tryptophan hydroxylase 2 (TPH2) mRNA, protein, and enzymatic activity throughout the male Sprague-Dawley rat brain. TPH2 is the genetic isoform of TPH that catalyzes the rate-limiting step in serotonin biosynthesis within the central nervous system. Although cell bodies of serotonergic neurons are located mainly in the raphe, serotonin-containing axons innervate many regions of the brain. In the present study, we assessed the levels of mRNA, protein expression, and enzyme activity of TPH2 in the rat raphe, ventral tegmental area (VTA), substantia nigra, hippocampus, cerebellum, dorsal striatum, nucleus accumbens, amygdala, and medial prefrontal cortex to more fully understand the distribution of this enzyme throughout the central nervous system. The pineal gland was used as a control tissue that expresses TPH1 (the peripheral enzyme), but not TPH2. As expected, the raphe showed the highest brain TPH2 activity and protein expression. In the contrast to other reports, however, the VTA followed the raphe as the region with the second-highest amount of TPH2 activity, mRNA and protein expression. There were significantly lower TPH activities and levels of TPH2 protein in the other regions. In addition, TPH2 immunocytochemistry demonstrated the presence of TPH-positive cell bodies within the VTA. The results of this study indicate that TPH2 and serotonergic signaling may play an important role in the mesolimbic/mesocortical reward pathway.