EMAIL THIS PAGE TO A FRIEND
Drug metabolism and pharmacokinetics
Regional distribution of drug-metabolizing enzyme activities in the liver and small intestine of cynomolgus monkeys.
PMID 21273732
Abstract
The cynomolgus monkey is an animal species widely used to study drug metabolism because of its evolutionary closeness to humans. However, drug-metabolizing enzyme activities have not been compared in various parts of the liver and small intestine in cynomolgus monkeys. In this study, therefore, drug-metabolizing enzyme activities were analyzed in the liver (the five lobes) and small intestine (six sections from the duodenum to the distal ileum). 7-Ethoxyresorufin O-deethylation, coumarin 7-hydroxylation, paclitaxel 6α-hydroxylation, diclofenac 4'-hydroxylation, tolbutamide methylhydroxylation, S-mephenytoin 4'-hydroxylation, bufuralol 1'-hydroxylation, chlorzoxazone 6-hydroxylation, midazolam 1'-hydroxylation, and testosterone 6β-, 16α-, 16β-, and 2α-hydroxylation were used as the probe reactions for this investigation. In liver, all probe reactions were detected and enzyme activity levels were similar in all lobes, whereas, in the small intestine, all enzyme activities were detected (except for coumarin 7-hydroxylase and testosterone 16α-hydroxylase activity), but from jejunum to ileum there was a decrease in the level of enzyme activity. This includes midazolam 1'-hydroxylation and testosterone 6β-hydroxylation, which are catalyzed by cynomolgus monkey cytochrome P450 (CYP) 3A4/5, orthologs of human CYP3A4/5, which are important drug-metabolizing enzymes. The data presented in this study are expected to facilitate the use of cynomolgus monkeys in drug metabolism studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
UC177
(±)-Mephenytoin
C12H14N2O2