EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Effect of concentration of trimethylchlorosilane (TMCS) and hexamethyldisilazane (HMDZ) silylating agents on surface free energy of silica aerogels.
PMID 21277584
Abstract
The surface free energy of a solid determines its surface and interfacial behavior in processes like wetting and adhesion which is crucial for silica aerogels in case of organic liquid absorption and transportation of chemicals at nano-scale for biotechnological applications. Here, we have demonstrated that the surface free energy of aerogels can be tuned in wide range from 5.5892 to 0.3073 mJ/m(2) by modifying their surface using TMCS and HMDZ silylating reagents. The alcogels were prepared by two step acid-base catalyzed process where the molar ratio of precursors Tetraethoxysilane (TEOS):Methanol (MeOH):Oxalic acid:NH(4)OH:NH(4)F was kept at optimal value of 1:2.7:0.18×10(-4):0.02:0.22×10(-3), respectively. To modify gel surfaces, TMCS and HMDZ concentration have been varied from 3% to 12% and such alcogels were dried at ambient pressure. It is observed from FTIR for aerogels that increase in concentration of silylating reagent resulted increase in hydrophobicity. This leads to increase in contact angle for water from 123° to 155° but leads to decrease in surface free energy from 5.5892 to 0.3073 mJ/m(2). As there is not direct method, we have used Neumann's equation of state to estimate surface energy of aerogels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
338869
Ammonium fluoride, ≥99.99% trace metals basis
H4FN
216011
Ammonium fluoride, ACS reagent, ≥98.0%
H4FN
224820
Ammonium hydrogen difluoride, reagent grade, 95%
H5F2N
324620
Lithium bis(trimethylsilyl)amide, 97%
C6H18LiNSi2
225770
Lithium bis(trimethylsilyl)amide solution, 1.0 M in THF
C6H18LiNSi2
577928
Lithium bis(trimethylsilyl)amide solution, 1 M in toluene
C6H18LiNSi2
766917
Lithium bis(trimethylsilyl)amide solution, 1.5 M in THF
C6H18LiNSi2
224367
Lithium bis(trimethylsilyl)amide solution, 1.0 M in hexanes
C6H18LiNSi2
577014
Lithium bis(trimethylsilyl)amide solution, 1 M in tert-butyl methyl ether
C6H18LiNSi2
775053
Lithium bis(trimethylsilyl)amide solution, 0.5 M in 2-methyltetrahydrofuran
C6H18LiNSi2
771864
Lithium bis(trimethylsilyl)amide solution, 0.5 M in toluene
C6H18LiNSi2
324671
Potassium bis(trimethylsilyl)amide, 95%
C6H18KNSi2
702722
Potassium bis(trimethylsilyl)amide solution, 1 M in THF
C6H18KNSi2
277304
Potassium bis(trimethylsilyl)amide solution, 0.5 M in toluene
C6H18KNSi2
775061
Potassium bis(trimethylsilyl)amide solution, 1.0 M in 2-methyltetrahydrofuran
C6H18KNSi2
766941
Potassium bis(trimethylsilyl)amide solution, 1.0 M in methyl tert-butyl ether
C6H18KNSi2
235083
Sodium bis(trimethylsilyl)amide, 95%
C6H18NNaSi2
245585
Sodium bis(trimethylsilyl)amide solution, 1.0 M in THF
C6H18NNaSi2
80631
Sodium bis(trimethylsilyl)amide solution, produced by Wacker Chemie AG, Burghausen, Germany, 40% in THF
C6H18NNaSi2
368059
Sodium bis(trimethylsilyl)amide solution, 0.6 M in toluene
C6H18NNaSi2
771546
Sodium bis(trimethylsilyl)amide solution, 0.5 M in methyl tert-butyl ether
C6H18NNaSi2
547816
Tris[N,N-bis(trimethylsilyl)amide]europium(III), 98%
C18H54EuN3Si6
547824
Tris[N,N-Bis(trimethylsilyl)amide]gadolinium(III), 98%
C18H54GdN3Si6
547980
Tris[N,N-bis(trimethylsilyl)amide]samarium(III), 98%
C18H54N3Si6Sm
524514
Tris[N,N-bis(trimethylsilyl)amide]yttrium
C18H54N3Si6Y