EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Automated precolumn derivatization of amino acids, small peptides, brain amines and drugs with primary amino groups for reversed-phase high-performance liquid chromatography using naphthalenedialdehyde as the fluorogenic label.
PMID 2127935
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
70215
2,3-Naphthalenedicarboxaldehyde, suitable for fluorescence
C12H8O2