EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Redox-neutral indole annulation cascades.
PMID 21280625
Abstract
Aminobenzaldehydes react with indoles in an unprecedented cascade reaction. This acid-catalyzed redox-neutral annulation proceeds via a condensation/1,5-hydride shift/ring-closure sequence. Polycyclic azepinoindoles and related compounds are obtained in a single step with good to excellent yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9628
2-Aminobenzaldehyde, ≥98%
C7H7NO