EMAIL THIS PAGE TO A FRIEND
Journal of biomedical optics
Light scattering of human red blood cells during metabolic remodeling of the membrane.
PMID 21280900
Abstract
We present the light scattering properties of individual human red blood cells (RBCs). We show that both the RBC static and dynamic scattering signals are altered by adenosine 5'-triphosphate (ATP)-driven membrane metabolic remodeling. To measure the light scattering signal from individual RBCs, we use diffraction phase microscopy together with a Fourier transform light scattering technique. RBC cytosolic ATPs are both chemically and metabolically depleted, and the corresponding scattering signals are compared with the light scattering signal of normal RBCs having physiologic levels of ATP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3221
Iodoacetamide, Single use vial of 56 mg
C2H4INO