EMAIL THIS PAGE TO A FRIEND
Methods in cell biology
Flow cytometric measurement of intracellular ionized calcium in single cells with indo-1 and fluo-3.
PMID 2128108
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
57180
Indo 1, for fluorescence, ≥80.0% (HPLC)
C32H26K5N3O12