EMAIL THIS PAGE TO A FRIEND
Biological trace element research
Dynamic pattern of gene expression of ZnT-4, caspase-3, LC3, and PRG-3 in rat cerebral cortex following flurothyl-induced recurrent neonatal seizures.
PMID 21286846
Abstract
Zinc transporters, plasticity-related genes, and autophagic/apoptotic pathway both are associated with developmental seizure-induced brain excitotoxicity. Here, for the first time, we report the timing of expression pattern of zinc transporter 4 (ZnT-4), plasticity-related gene 3 (PRG-3), specific marker of autophagic vacuoles (LC3), and apoptotic marker caspase-3 in cerebral cortex following neonatal seizures. A seizure was induced by inhalant flurothyl daily in neonatal Sprague-Dawley rats from postnatal day 6 (P6). Rats were assigned into the recurrent-seizure group (RS, seizures induced in six consecutive days) and the control group. At 1.5 h, 3 h, 6 h, 12 h, 24 h, 48 h, 7 days, and 14 days after the last seizure, the mRNA level of the four genes in cerebral cortex was detected using RT-PCR method. At an early period 6 h or 12 h after the last seizures, both ZnT-4 and LC3 showed significantly up-regulated mRNA level while PRG-3 showed significantly down-regulated mRNA level at 12 h in cerebral cortex of RS group than those at the corresponding time point in control group. In the long-term time point of 7 days after the last seizure, the mRNA level of caspase-3 down-regulated; meanwhile, there was up-regulated mRNA level of LC-3 in RS group when compared to the control rats. This is the first report investigating the gene expression pattern of ZnT-4, PRG-3, LC-3, and caspase-3 in the developing brain. The results suggest that the disturbed expression pattern of the four genes might play a role in the pathophysiology of recurrent neonatal seizure-induced acute and long-term brain damage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
287571
Bis(2,2,2-trifluoroethyl) ether, 98%
C4H4F6O