EMAIL THIS PAGE TO A FRIEND
Biotechnology letters
Co-loading of bisphosphonates and antibiotics to a biomimetic hydroxyapatite coating.
PMID 21287232
Abstract
We have incorporated bisphosphonates and antibiotics simultaneously into a biomimetic hydroxyapatite implant coating aiming to use the interaction between drug-molecules and hydroxyapatite to enable local release of the two different substances to obtain a dual biological effect. A sustained release over for 43 h of antibiotics (cephalothin) was achieved without negative interference from the presence of the bisphosphonate (clodronate) which, in turn, successfully bonded to the coating surface. To our knowledge, this is the first study that indicates the possibility to simultaneously incorporate both antibiotics and bisphosphonates to an implant coating, a strategy that is believed to improve implant stability and reduce implant-related infections.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4520
Cephalothin sodium salt
C16H15N2NaO6S2