EMAIL THIS PAGE TO A FRIEND
American journal of health-system pharmacy : AJHP : official journal of the American Society of Health-System Pharmacists
Iloperidone: Does it have a meaningful place in therapy?
PMID 21289322