EMAIL THIS PAGE TO A FRIEND
Nanotechnology
Lithographical gap-size engineered nanoarrays for surface-enhanced Raman probing of biomarkers.
PMID 21289393
Abstract
Engineered gold nanostructured arrays with feature separation in the sub-10-nm range were fabricated and used for highly sensitive surface enhanced Raman scattering (SERS) detection of biomarkers. Nanostructuring is achieved by combining electron beam lithography with the so-called shadow evaporation technique which involves a two-step Au vapor deposition through a suspended Ge mask while the substrate is tilted in opposite directions. This results in a regular triangular surface pattern with extremely small gap distances that allow positive enhancement of the local electric fields by enabling improvements in the electromagnetic coupling between adjacent nanoparticles. The resulting SERS active surfaces are suitable for the realization of reference procedures for quantifying marker molecules like urea or creatinine at physiologically relevant concentrations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
662534
4-Mercaptobenzoic acid, technical grade, 90%
C7H6O2S
706329
4-Mercaptobenzoic acid, 99%
C7H6O2S