EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Diversity-oriented synthesis of polycyclic scaffolds by modification of an anodic product derived from 2,4-dimethylphenol.
PMID 21290525
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D174203
2,4-Dimethylphenol, 98%
C8H10O
W506206
2,4-Xylenol, ≥98%
C8H10O