EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
NMR structural study of fructans produced by Bacillus sp. 3B6, bacterium isolated in cloud water.
PMID 21292244
Abstract
Bacillus sp. 3B6, bacterium isolated from cloud water, was incubated on sucrose for exopolysaccharide production. Dialysis of the obtained mixture (MWCO 500) afforded dialyzate (DIM) and retentate (RIM). Both were separated by size exclusion chromatography. RIM afforded eight fractions: levan exopolysaccharide (EPS), fructooligosaccharides (FOSs) of levan and inulin types with different degrees of polymerization (dp 2-7) and monosaccharides fructose:glucose=9:1. Levan was composed of two components with molecular mass ~3500 and ~100kDa in the ratio 2.3:1. Disaccharide fraction contained difructose anhydride DFA IV. 1-Kestose, 6-kestose, and neokestose were identified as trisaccharides in the ratio 2:1:3. Fractions with dp 4-7 were mixtures of FOSs of levan (2,6-βFruf) and inulin (1,2-βFruf) type. DIM separation afforded two dominant fractions: monosaccharides with fructose: glucose ratio 1:3; disaccharide fraction contained sucrose only. DIM trisaccharide fraction contained 1-kestose, 6-kestose, and neokestose in the ratio1.5:1:2, penta and hexasaccharide fractions contained FOSs of levan type (2,6-βFruf) containing α-glucose. In the pentasaccharide fraction also the presence of a homopentasaccharide composed of 2,6-linked βFruf units only was identified. Nystose, inulin (1,2-βFruf) type, was identified as DIM tetrasaccharide. Identification of levan 2,6-βFruf and inulin 1,2-βFruf type oligosaccharides in the incubation medium suggests both levansucrase and inulosucrase enzymes activity in Bacillus sp. 3B6.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L8647 Levan from Erwinia herbicola