EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Determination of volatile organic compounds in recycled polyethylene terephthalate and high-density polyethylene by headspace solid phase microextraction gas chromatography mass spectrometry to evaluate the efficiency of recycling processes.
PMID 21292271
Abstract
A method for the determination of volatile organic compounds (VOCs) in recycled polyethylene terephthalate and high-density polyethylene using headspace sampling by solid-phase microextraction and gas chromatography coupled to mass spectrometry detection is presented. This method was used to evaluate the efficiency of cleaning processes for VOC removal from recycled PET. In addition, the method was also employed to evaluate the level of VOC contamination in multilayer packaging material containing recycled HDPE material. The optimisation of the extraction procedure for volatile compounds was performed and the best extraction conditions were found using a 75 μm carboxen-polydimethylsiloxane (CAR-PDMS) fibre for 20 min at 60 °C. The validation parameters for the established method were linear range, linearity, sensitivity, precision (repeatability), accuracy (recovery) and detection and quantification limits. The results indicated that the method could easily be used in quality control for the production of recycled PET and HDPE.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
57318 SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm, for use with manual holder, needle size 24 ga
57344-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (partially crosslinked phase), needle size 23 ga, for use with manual holder
57319 SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (partially crosslinked), needle size 24 ga, for use with autosampler
57343-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (partially crosslinked phase), needle size 23 ga, for use with autosampler
57334-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 24 ga, for use with manual holder, StableFlex fiber
57335-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 24 ga, StableFlex, for use with autosampler
57295-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 23 ga, StableFlex, for use with autosampler
57906-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm (bonded phase), for use with autosampler, needle size 23 ga, metal alloy fiber
FFA57295-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 23 ga, Stableflex, for use with multi fiber exchanger
FFA57335-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, for use with multi fiber exchanger, needle size 24 ga, StableFlex
504831 SPME Portable Field Sampler, coating Carboxen/Polydimethylsiloxane (CAR/PDMS)