Journal of agricultural and food chemistry
Extracts of Maqui ( Aristotelia chilensis ) and Murta ( Ugni molinae Turcz.): sources of antioxidant compounds and α-Glucosidase/α-Amylase inhibitors.
PMID 21294510
The objective of this work was to evaluate the antioxidant and antihemolytic activities of crude, aqueous, and organic-aqueous extracts of maqui ( Aristotelia chilensis ) and murta ( Ugni molinae Turcz.), together with their inhibiting effect on enzymes involved in the metabolism of carbohydrates. Radical scavenging activity, inhibition of linoleic acid oxidation in a micellar system, antihemolytic activity, and inhibition of α-amylases and α-glucosidases were analyzed. Crude extracts of maqui leaves and fruits were found to be important sources of polyphenolic compounds, showing 69.0 ± 0.9 and 45.7 ± 1.1 mg GAE/g dm, respectively. Polyphenols from maqui leaves were active as antioxidants and antihemolytic compounds (p < 0.05), showing a noncompetitive inhibiting effect on α-glucosidase. Flavan-3-ol polymers and glycosylated flavonols, such as quercetin glucoside and kaempferol glucoside, were tentatively identified in extracts. This preliminary observation provides the basis for further examination of the suitability of polyphenol-enriched extracts from maqui and murta as nutritional or medicinal supplements with potential human health benefits.