EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Development of a high throughput screen for allosteric modulators of melanocortin-4 receptor signaling using a real time cAMP assay.
PMID 21296065
Abstract
The melanocortin MC(4) receptor is a potential target for the development of drugs for both obesity and cachexia. Melanocortin MC(4) receptor ligands known thus far are orthosteric agonists or antagonists, however the agonists, in particular, have generally exhibited unwanted side effects. For some receptors, allosteric modulators are expected to reduce side-effect profiles. To identify allosteric modulators of the melanocortin MC(4) receptor, we created HEK293 cell lines coexpressing the human melanocortin MC(4) receptor and a modified luciferase-based cAMP sensor. Monitoring luminescence as a readout of real-time intracellular cAMP concentration, we demonstrate that this cell line is able to report melanocortin agonist responses, as well as inverse agonist response to the physiological AgRP peptide. Based on the MC4R-GLO cell line, we developed an assay that was shown to meet HTS standards (Z'=0.50). A pilot screen run on the Microsource Spectrum compound library (n=2000) successfully identified 62 positive modulators. This screen identified predicted families of compounds: β(2)AR agonists - the β(2)AR being endogenously expressed in HEK293 cells, an adenylyl cyclase activator and finally a distribution of phosphodiesterase (PDE) inhibitors well characterized or recently identified. In this last category, we identified a structural family of coumarin-derived compounds (imperatorin, osthol and prenyletin), along with deracoxib, a drug in veterinary use for its COX2 inhibitory properties. This latter finding unveiled a new off-target mechanism of action for deracoxib as a PDE inhibitor. Overall, these data are the first report of a HTS for allosteric modulators for a Gs protein coupled receptor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML2014 Deracoxib, ≥98% (HPLC)
C17H14F3N3O3S
I6659
Imperatorin, ≥98% (HPLC), powder
C16H14O4