EMAIL THIS PAGE TO A FRIEND
Cancer letters
The tSP Motif in aP180 inhibits phospholipase d1 activity resulting in increased efficacy of anticancer drug via its direct binding to carboxyl terminal of phospholipase d1.
PMID 21296491