EMAIL THIS PAGE TO A FRIEND
Indian journal of experimental biology
Co-polymeric hydrophilic nanospheres for drug delivery: release kinetics, and cellular uptake.
PMID 21299047
Abstract
Nanobiotechnology focuses on the biological effects and applications of nanoparticles that include nano-safety, drug encapsulation and nanotherapeutics. The present study focuses on hydrophilic nanospheres of copolymers N-isopropylacrylamide [NIPAAM] and vinyl pyrrolidone [VP], encapsulating a bioactive derivative of 5-fluorouracil-hexyl-carbamoyl fluorouracil (HCFU). The size of the nanospheres was approximately 58 nm and the surface charge measured was -15.4 mV. Under optimal conditions, the yield was >80%, and the drug loading was 2%. The entrapment efficiency was approximately 75%. Wide-angle X-ray diffraction analysis showed that the entrapped HCFU was present in an amorphous state, which has higher water solubility compared with the crystalline state. Slow drug release from nanospheres was observed in PBS and serum, with approximately 80% released at 37 degrees C after 72 h. The HCFU loaded polymeric nanospheres have been found to be stable in whole blood having negligible RBC toxicity. Cytotoxicity in Mia-Paca 3, pancreatic cancer cell line was done in a 24-72 h assay. Dose dependant cytotoxicity was observed when incubated with various concentrations of HCFU loaded polymeric nanospheres while HCFU per se (<1 mg) showed 90% toxicity within 24 h.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1494
Carmofur, ≥98% (HPLC), powder
C11H16FN3O3