EMAIL THIS PAGE TO A FRIEND
Journal of the science of food and agriculture
Schisandrin enhances dendrite outgrowth and synaptogenesis in primary cultured hippocampal neurons.
PMID 21302324
Abstract
Schisandra chinensis, commonly used in Asia for tea material and traditional Chinese medicine, is presumed to enhance mental and intellectual functions. In this study, the effects and signalling mechanisms of a purified compound schisandrin, one of the lignan of Schisandra chinensis, on primary cultured hippocampal neurons were investigated. Schisandrin treatment enhanced total dendritic length and branching complexity, both of which were significantly suppressed in the presence of specific blockers for calmodulin-dependent kinase II (CaMKII), protein kinase C epsilon (PKCε), and mitogen activated protein kinase kinase (MEK). Moreover, schisandrin induced calcium influx, and phosphorylation of CaMKII, PKCε, and MEK. Inhibition of CAMKII and PKCε attenuated the schisandrin-induced phosphorylation of PKCε and MEK, and the phosphorylation of MEK, respectively. Moreover, schisandrin also stimulated the phosphorylation of cyclic AMP responsive-element binding protein (CREB) at Ser-133, an effect that was blocked by KN93. In addition to its neuritogenic effects, schisandrin increased the numbers of postsynaptic density-95-positive and FM1-43-positive puncta in dendrites and synaptic boutons, respectively. In hippocampal neurons, schisandrin exhibits neurotrophic properties that are mediated by the CaMKII-PKCε-MEK pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0054
Schizandrin, ≥98% (HPLC)
C24H32O7