EMAIL THIS PAGE TO A FRIEND
The Journal of chemical physics
Bond dissociation of the dipeptide dialanine and its derivative alanine anhydride induced by low energy electrons.
PMID 21303118
Abstract
Dissociative electron attachment to dialanine and alanine anhydride has been studied in the gas phase utilizing a double focusing two sector field mass spectrometer. We show that low-energy electrons (i.e., electrons with kinetic energies from near zero up to 13 eV) attach to these molecules and subsequently dissociate to form a number of anionic fragments. Anion efficiency curves are recorded for the most abundant anions by measuring the ion yield as a function of the incident electron energy. The present experiments show that as for single amino acids (M), e.g., glycine, alanine, valine, and proline, the dehydrogenated closed shell anion (M-H)(-) is the most dominant reaction product. The interpretation of the experiments is aided by quantum chemical calculations based on density functional theory, by which the electrostatic potential and molecular orbitals are calculated and the initial electron attachment process prior to dissociation is investigated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9502
Ala-Ala
C6H12N2O3
362557
Alanine anhydride, 99%
C6H10N2O2
A0912
D-Ala-D-Ala
C6H12N2O3
A9377
DL-Ala-DL-Ala
C6H12N2O3