EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Pd-NHC catalyzed conjugate addition versus the Mizoroki-Heck reaction.
PMID 21305628
Abstract
Ace of base: A catalytic system is presented that, solely by choice of the base, selectively switches between conjugate addition and the Mizoroki-Heck reaction of aryl halides with Michael acceptors (see scheme; R, R' = alkyl, aryl). For conjugate addition reactions, this avoids the preparation and use of organometallics.
Related Materials