EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Rhodium-catalyzed enantioselective addition of boronic acids to N-benzylnicotinate salts.
PMID 21309573
Abstract
The highly enantioselective catalytic asymmetric addition of aryl and alkenylboronic acids to N-benzylnicotinate salt 1 is described. The dihydropyridine 2 reaction products can be converted to synthetically useful piperidines. Application of the methodology to the preparation of enantioenriched quaternary chiral centers is also discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72351
Benzyl nicotinate, ≥98.0% (GC)
C13H11NO2